siRNA Details
| Gene Symbol: Lyrm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001076681.2 | NP_001070149.1 | 3 | 254 | ||
| GenBank mRNA | AK007467.1 | - | 3 | 268 | ||
| AK009070.1 | - | 3 | 237 | |||
| AK144380.1 | - | 3 | 255 | |||
| BC130224.1 | - | 3 | 245 | 40141726 | ||

Lyrm9
Genomic Map