siRNA Details
Gene Symbol: Tmem128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025480.3 | NP_079756.2 | 3 | 324 | ||
GenBank mRNA | AK012789.2 | - | 3 | 325 | ||
AK013227.1 | - | 3 | 324 | |||
AK076679.1 | - | 3 | 316 | |||
AK080853.1 | - | 3 | 325 | |||
AK162257.1 | - | 3 | 319 | |||
BC025197.1 | - | 3 | 483 | 5251991 | ||
BC060969.1 | - | 3 | 313 | 6704414 | ||
BC110310.1 | - | 3 | 307 | 5325405 |
