siRNA Details
| Gene Symbol: Ccdc82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025534.2 | NP_079810.2 | 7 | 1291 | ||
| GenBank mRNA | AK088611.1 | - | 2 | 258 | ||
| BC040405.1 | - | 5 | 452 | |||
| BC057326.1 | - | 7 | 1291 | 6826080 | ||
| BC098496.1 | - | 6 | 1218 | 30089818 | ||

Ccdc82
Genomic Map