siRNA Details
| Gene Symbol: Mcts2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025543.2 | NP_079819.1 | 1 | 652 | ||
| GenBank mRNA | AK002563.1 | - | 1 | 630 | ||
| AK010255.1 | - | 1 | 653 | |||
| AK028001.1 | - | 1 | 631 | |||
| BC027507.1 | - | 1 | 645 | 1478068 | ||

Mcts2
Genomic Map