siRNA Details
| Gene Symbol: Pcp4l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025557.1 | NP_079833.1 | 1, 2 | 256 | ||
| GenBank mRNA | AK002772.1 | - | 1, 2 | 227 | ||
| AK032164.1 | - | 1, 2 | 226 | |||
| AK075738.1 | - | 1, 2 | 226 | |||
| AY304481.1 | - | 1, 2 | 227 | |||
| BC116208.1 | - | 1, 2 | 286 | |||
| BC126965.1 | - | 1, 2 | 286 | |||
| BY238909.1 | - | 1, 2 | 256 | |||

Pcp4l1
Genomic Map