siRNA Details
| Gene Symbol: Pcp4l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025557.1 | NP_079833.1 | 3 | 356 | ||
| GenBank mRNA | AK002772.1 | - | 3 | 327 | ||
| AK032164.1 | - | 3 | 326 | |||
| AK075738.1 | - | 3 | 326 | |||
| AY304481.1 | - | 3 | 327 | |||
| BC116208.1 | - | 3 | 386 | |||
| BC126965.1 | - | 3 | 386 | |||
| BY238909.1 | - | 3 | 356 | |||

Pcp4l1
Genomic Map