siRNA Details
| Gene Symbol: Exosc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081188.1 | NP_001074657.1 | 7 | 672 | ||
| GenBank mRNA | AK004437.1 | - | 4 | 244 | ||
| AK011294.1 | - | 7 | 659 | |||
| AK129057.1 | - | 7 | 624 | |||
| BC039655.1 | - | 7 | 657 | |||
| BC094932.1 | - | 7 | 665 | 30008071 | ||
| BC132363.1 | - | 7 | 687 | 40130640 | ||
| BC145675.1 | - | 7 | 687 | 40130978 | ||

Exosc7
Genomic Map