siRNA Details
| Gene Symbol: Exosc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081188.1 | NP_001074657.1 | 5 | 471 | ||
| GenBank mRNA | AK004437.1 | - | 2 | 43 | ||
| AK011294.1 | - | 5 | 458 | |||
| AK129057.1 | - | 5 | 423 | |||
| BC039655.1 | - | 5 | 456 | |||
| BC052656.1 | - | 5 | 453 | 30041585 | ||
| BC094932.1 | - | 5 | 464 | 30008071 | ||
| BC132363.1 | - | 5 | 486 | 40130640 | ||
| BC145675.1 | - | 5 | 486 | 40130978 | ||

Exosc7
Genomic Map