siRNA Details
| Gene Symbol: Fam107b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025626.4 | NP_079902.1 | 2 | 341 | ||
| GenBank mRNA | AK013932.2 | - | 2 | 306 | ||
| AK019815.1 | - | 2 | 253 | |||
| AK032875.1 | - | 2 | 342 | |||
| AK168759.1 | - | 2 | 307 | |||
| BC021353.1 | - | 2 | 307 | 5006672 | ||

Fam107b
Genomic Map