siRNA Details
| Gene Symbol: Ankrd61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025732.3 | NP_080008.1 | 2 | 341 | ||
| GenBank mRNA | AK014929.1 | - | 2 | 342 | ||
| AK017053.1 | - | 2 | 282 | |||
| BC052346.1 | - | 2 | 563 | 6703467 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd61| Gene Symbol: Ankrd61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025732.3 | NP_080008.1 | 2 | 341 | ||
| GenBank mRNA | AK014929.1 | - | 2 | 342 | ||
| AK017053.1 | - | 2 | 282 | |||
| BC052346.1 | - | 2 | 563 | 6703467 | ||