siRNA Details
| Gene Symbol: Ankrd61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025732.3 | NP_080008.1 | 3 | 660 | ||
| GenBank mRNA | AK014929.1 | - | 3 | 661 | ||
| AK017053.1 | - | 3 | 601 | |||
| AK146023.1 | - | 939 | ||||
| BC052346.1 | - | 3 | 882 | 6703467 | ||

Ankrd61
Genomic Map