siRNA Details
| Gene Symbol: 4933436I01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025763.1 | NP_080039.1 | 2 | 1043 | ||
| GenBank mRNA | AK017084.1 | - | 2 | 1043 | ||
| BC125317.1 | - | 2 | 1029 | 40129832 | ||
| BC138016.1 | - | 2 | 1029 | 8861035 | ||

4933436I01Rik
Genomic Map