siRNA Details
| Gene Symbol: Zfp869 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039965.1 | NP_001035054.1 | 4 | 603 | ||
| NM_001039967.1 | NP_001035056.1 | 5 | 711 | |||
| NM_181274.3 | NP_851791.1 | 4 | 671 | |||
| GenBank mRNA | AK004570.1 | - | 4 | 614 | ||
| AK030679.1 | - | 5 | 709 | |||
| AK083392.1 | - | 5 | 626 | |||
| AK136878.1 | - | 4 | 607 | |||
| BC063330.1 | - | 668 | ||||
| BC079868.1 | - | 6 | 1206 | |||

Zfp869
Genomic Map