siRNA Details
| Gene Symbol: Acad8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025862.2 | NP_080138.2 | 3 | 321 | ||
| GenBank mRNA | AK009379.1 | - | 3 | 300 | ||
| AK019502.1 | - | 3 | 318 | |||
| AK029897.1 | - | 3 | 285 | |||
| AK051091.1 | - | 3 | 276 | |||
| AK077335.1 | - | 3 | 301 | |||
| AK161255.1 | - | 3 | 275 | |||
| AK211554.1 | - | 1 | 40 | |||
| BC026535.1 | - | 3 | 295 | |||
| BC037644.1 | - | 3 | 297 | 5053565 | ||
| BQ931973.1 | - | 3 | 321 | |||

Acad8
Genomic Map