siRNA Details
| Gene Symbol: Mrps18b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025878.1 | NP_080154.1 | 4 | 336 | ||
| GenBank mRNA | AB049954.1 | - | 4 | 360 | ||
| AK010250.1 | - | 4 | 343 | |||
| AK088540.1 | - | 4 | 332 | |||
| AK166597.1 | - | 4 | 357 | |||
| AK167067.1 | - | 4 | 337 | |||
| AK168140.1 | - | 4 | 352 | |||
| BC021752.1 | - | 4 | 336 | 5136720 | ||

Mrps18b
Genomic Map