siRNA Details
| Gene Symbol: Mrps18b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025878.1 | NP_080154.1 | 1 | 55 | ||
| GenBank mRNA | AB049954.1 | - | 1 | 79 | ||
| AK002554.1 | - | 1 | 76 | |||
| AK076033.1 | - | 1 | 76 | |||
| AK076044.1 | - | 1 | 76 | |||
| AK166597.1 | - | 1 | 76 | |||
| AK167067.1 | - | 1 | 56 | |||
| AK168140.1 | - | 1 | 71 | |||
| BC021752.1 | - | 1 | 55 | 5136720 | ||

Mrps18b
Genomic Map