siRNA Details
Gene Symbol: 2610002M06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025921.3 | NP_080197.2 | 1, 2 | 196 | ||
GenBank mRNA | AK011297.1 | - | 1, 2 | 144 | ||
AK019383.1 | - | 1, 2 | 17 | |||
BC016070.1 | - | 1, 2 | 135 | 4481241 |
