siRNA Details
| Gene Symbol: Fancl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277273.1 | NP_001264202.1 | 9 | 870 | ||
| NM_025923.3 | NP_080199.1 | 9 | 870 | |||
| GenBank mRNA | AF513619.1 | - | 9 | 870 | ||
| AK008605.1 | - | 9 | 756 | |||
| AK011520.1 | - | 9 | 775 | |||
| AK011752.1 | - | 9 | 747 | |||
| AK011903.1 | - | 9 | 753 | |||
| AK018018.1 | - | 9 | 752 | |||
| AK020981.1 | - | 4 | 324 | |||
| AK076017.1 | - | 9 | 753 | |||
| AK168538.1 | - | 9 | 754 | |||
| BC032876.1 | - | 9 | 744 | 1282316 | ||
| BC066181.1 | - | 9 | 799 | 6435171 | ||
| CN455644.1 | - | 7 | 505 | |||

Fancl
Genomic Map