siRNA Details
| Gene Symbol: Ceacam14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025957.4 | NP_080233.1 | 2 | 413 | ||
| GenBank mRNA | AK005390.1 | - | 2 | 414 | ||
| AK005482.1 | - | 2 | 414 | |||
| AK005533.1 | - | 2 | 414 | |||
| BC099412.1 | - | 2 | 343 | 30787387 | ||

Ceacam14
Genomic Map