siRNA Details
| Gene Symbol: Fbxo5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025995.2 | NP_080271.2 | 2 | 300 | ||
| GenBank mRNA | AK011820.1 | - | 2 | 298 | ||
| AK135916.1 | - | 2 | 255 | |||
| AK160201.1 | - | 2 | 256 | |||
| BC053434.1 | - | 2 | 224 | 6336178 | ||

Fbxo5
Genomic Map