siRNA Details
| Gene Symbol: Nkain1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025998.3 | NP_080274.2 | 4 | 578 | ||
| GenBank mRNA | AK011164.1 | - | 4 | 711 | ||
| AK011850.1 | - | 4 | 509 | |||
| AK076029.1 | - | 1 | 66 | |||
| AK184838.1 | - | 1 | 31 | |||
| AK187454.1 | - | 1 | 31 | |||
| AK205247.1 | - | 1 | 31 | |||
| AK205326.1 | - | 1 | 31 | |||
| AK205517.1 | - | 1 | 31 | |||
| AK206217.1 | - | 1 | 31 | |||
| AK206243.1 | - | 1 | 31 | |||
| AK208705.1 | - | 1 | 31 | |||
| AK208715.1 | - | 1 | 31 | |||
| AK208816.1 | - | 1 | 31 | |||
| AK209134.1 | - | 1 | 31 | |||
| AK209985.1 | - | 1 | 31 | |||
| AK210092.1 | - | 1 | 31 | |||
| AK210264.1 | - | 1 | 31 | |||
| AK211065.1 | - | 1 | 31 | |||
| AK211305.1 | - | 1 | 31 | |||
| AK211393.1 | - | 1 | 31 | |||
| AK211541.1 | - | 1 | 31 | |||
| AK213620.1 | - | 1 | 31 | |||
| AK214155.1 | - | 1 | 31 | |||
| AK215330.1 | - | 1 | 31 | |||
| AK216558.1 | - | 1 | 31 | |||
| AK216625.1 | - | 1 | 31 | |||
| AK216635.1 | - | 1 | 31 | |||
| AK216972.1 | - | 1 | 31 | |||
| AK217347.1 | - | 1 | 31 | |||
| AK218646.1 | - | 1 | 31 | |||
| AK218997.1 | - | 1 | 31 | |||
| AK219655.1 | - | 1 | 31 | |||
| AK220029.1 | - | 1 | 31 | |||
| BC116391.1 | - | 4 | 508 | 40101865 | ||
| BC116392.1 | - | 4 | 525 | |||
| EF058048.2 | - | 4 | 631 | |||

Nkain1
Genomic Map