siRNA Details
| Gene Symbol: Dph7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026044.3 | NP_080320.1 | 9 | 1435 | ||
| GenBank mRNA | AK013297.1 | - | 9 | 1415 | ||
| AK078448.1 | - | 9 | 1398 | |||
| AK087998.1 | - | 8 | 1310 | |||
| AK152000.1 | - | 8 | 1348 | |||
| AK153537.1 | - | 8 | 1348 | |||
| BC056962.1 | - | 7 | 2085 | |||
| BC086486.1 | - | 9 | 1437 | |||

Dph7
Genomic Map