siRNA Details
| Gene Symbol: Ccdc53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122960.1 | NP_001116432.1 | 5 | 503 | ||
| NM_026070.3 | NP_080346.1 | 5 | 506 | |||
| NM_027487.1 | NP_081763.1 | 5 | 506 | |||
| GenBank mRNA | AK004206.1 | - | 5 | 421 | ||
| AK009358.1 | - | 5 | 402 | |||
| AK013835.1 | - | 5 | 511 | |||
| AK017732.1 | - | 5 | 434 | |||
| AK158822.1 | - | 5 | 507 | |||
| BC029256.1 | - | 5 | 403 | 3661472 | ||
| CK795584.1 | - | 5 | 455 | |||

Ccdc53
Genomic Map