siRNA Details
| Gene Symbol: Calcoco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026192.3 | NP_080468.1 | 6, 7 | 918 | ||
| GenBank mRNA | AK007393.1 | - | 6, 7 | 809 | ||
| AK018734.2 | - | 6, 7 | 804 | |||
| AK035598.1 | - | 6, 7 | 830 | |||
| AK087513.1 | - | 6, 7 | 872 | |||
| AK207562.1 | - | 1, 2 | 144 | |||
| AK217037.1 | - | 1, 2 | 144 | |||
| AK220400.1 | - | 6, 7 | 831 | |||
| AY131329.1 | - | 5, 6 | 784 | |||
| BC054783.1 | - | 6, 7 | 812 | 6815943 | ||

Calcoco1
Genomic Map