siRNA Details
| Gene Symbol: Calcoco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026192.3 | NP_080468.1 | 13 | 1869 | ||
| GenBank mRNA | AK007393.1 | - | 13 | 1760 | ||
| AK018734.2 | - | 13 | 1756 | |||
| AK035598.1 | - | 13 | 1781 | |||
| AK087513.1 | - | 13 | 1823 | |||
| AK220400.1 | - | 13 | 1782 | |||
| AY131329.1 | - | 12 | 1735 | |||
| BC054783.1 | - | 13 | 1763 | 6815943 | ||

Calcoco1
Genomic Map