siRNA Details
| Gene Symbol: Bend5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026279.3 | NP_080555.1 | 1, 2 | 401 | ||
| GenBank mRNA | AK014057.1 | - | 1, 2 | 391 | ||
| AK075885.1 | - | 1, 2 | 401 | |||
| AK156708.1 | - | 1, 2 | 221 | |||
| AK167425.1 | - | 1, 2 | 221 | |||
| BC037633.1 | - | 1, 2 | 234 | |||

Bend5
Genomic Map