siRNA Details
| Gene Symbol: Anp32b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130889.2 | NP_570959.1 | 1, 2 | 241 | ||
| GenBank mRNA | AB025582.1 | - | 1, 2 | 62 | ||
| AK011134.1 | - | 1, 2 | 373 | |||
| AK136648.1 | - | 1, 2 | 75 | |||
| AK165807.1 | - | 1, 2 | 311 | |||
| BC003489.1 | - | 1, 2 | 259 | 2811622 | ||
| BC005628.1 | - | 1, 2 | 267 | 3963541 | ||
| BC093506.1 | - | 1, 2 | 231 | 4917194 | ||

Anp32b
Genomic Map