siRNA Details
| Gene Symbol: Plgrkt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026362.2 | NP_080638.1 | 4 | 348 | ||
| GenBank mRNA | AK017171.1 | - | 4 | 372 | ||
| AK017215.1 | - | 4 | 349 | |||
| AK168454.1 | - | 4 | 324 | |||
| AK170700.1 | - | 5 | 422 | |||
| AK211954.1 | - | 3 | 286 | |||
| AK219906.1 | - | 3 | 286 | |||
| BC024953.1 | - | 4 | 307 | 4951101 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001106347, XM_006231203, XM_006231205, XM_008760323, XM_006231204.

Plgrkt
Genomic Map