siRNA Details
| Gene Symbol: Rmdn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033136.3 | NP_001028308.1 | 10, 11 | 1372 | ||
| GenBank mRNA | AK004786.1 | - | 10, 11 | 1298 | ||
| AK146299.1 | - | 10, 11 | 1291 | |||
| AK151090.1 | - | 10, 11 | 1293 | |||
| AK153322.1 | - | 10, 11 | 1293 | |||
| AK153369.1 | - | 10, 11 | 1292 | |||
| AK171770.1 | - | 10, 11 | 1373 | |||
| BC055754.1 | - | 10, 11 | 1266 | |||

Rmdn3
Genomic Map