siRNA Details
Gene Symbol: Sec14l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144520.2 | NP_653103.1 | 10 | 928 | ||
GenBank mRNA | BC005759.1 | - | 10 | 928 | 2655022 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sec14l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144520.2 | NP_653103.1 | 10 | 928 | ||
GenBank mRNA | BC005759.1 | - | 10 | 928 | 2655022 |