siRNA Details
| Gene Symbol: Cdca5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026410.3 | NP_080686.1 | 6 | 811 | ||
| GenBank mRNA | AK010540.1 | - | 6 | 778 | ||
| AK011701.1 | - | 6 | 811 | |||
| AK016305.1 | - | 6 | 925 | |||
| BC029626.1 | - | 5 | 705 | |||
| BC052904.1 | - | 6 | 809 | 30065848 | ||

Cdca5
Genomic Map