siRNA Details
| Gene Symbol: Ddx59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026500.3 | NP_080776.1 | 2 | 947 | ||
| GenBank mRNA | AK004833.1 | - | 2 | 948 | ||
| AK013179.1 | - | 2 | 909 | |||
| AK076462.1 | - | 2 | 902 | |||
| AK133731.1 | - | 2 | 909 | |||
| BC023840.1 | - | 2 | 874 | 5344890 | ||

Ddx59
Genomic Map