siRNA Details
| Gene Symbol: Chac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290667.1 | NP_001277596.1 | 1 | 176 | ||
| NM_026527.3 | NP_080803.1 | 1 | 176 | |||
| GenBank mRNA | AK007629.1 | - | 1 | 100 | ||
| AK010914.1 | - | 1 | 146 | |||
| AK021391.1 | - | 1 | 146 | |||
| AK078993.1 | - | 1 | 162 | |||
| BC025100.1 | - | 1 | 90 | 3967723 | ||
| BY333527.1 | - | 1 | 176 | |||

Chac2
Genomic Map