siRNA Details
| Gene Symbol: Chac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290667.1 | NP_001277596.1 | 2 | 493 | ||
| NM_026527.3 | NP_080803.1 | 3 | 529 | |||
| GenBank mRNA | AK007629.1 | - | 3 | 453 | ||
| AK010914.1 | - | 3 | 499 | |||
| AK021391.1 | - | 2 | 463 | |||
| AK078993.1 | - | 3 | 515 | |||
| BC025100.1 | - | 3 | 443 | 3967723 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001025016, XM_008770464, BC082031.

Chac2
Genomic Map