siRNA Details
| Gene Symbol: Mpnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026530.5 | NP_080806.4 | 11 | 1283 | ||
| GenBank mRNA | AK012128.1 | - | 10 | 1238 | ||
| AK146973.1 | - | 11 | 1268 | |||
| AK160356.1 | - | 11 | 1252 | |||
| AK172381.1 | - | 10 | 1026 | |||
| BC017625.1 | - | 10 | 1232 | |||

Mpnd
Genomic Map