siRNA Details
Gene Symbol: Fam173b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026546.3 | NP_080822.1 | 2 | 200 | ||
GenBank mRNA | AK020868.1 | - | 2 | 200 | ||
BC030179.1 | - | 1 | 191 | 5039149 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam173b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026546.3 | NP_080822.1 | 2 | 200 | ||
GenBank mRNA | AK020868.1 | - | 2 | 200 | ||
BC030179.1 | - | 1 | 191 | 5039149 |