siRNA Details
| Gene Symbol: Fam92a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026558.4 | NP_080834.3 | 7, 8 | 939 | ||
| GenBank mRNA | AK013129.1 | - | 6, 7 | 768 | ||
| AK019574.1 | - | 7, 8 | 767 | |||
| AK078438.1 | - | 7, 8 | 920 | |||
| AK088705.1 | - | 7, 8 | 920 | |||
| AK214863.1 | - | 1, 2 | 14 | |||
| BC020162.1 | - | 733 | 3991990 | |||
| BC055848.1 | - | 7, 8 | 1857 | |||

Fam92a
Genomic Map