siRNA Details
| Gene Symbol: Hyi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026601.3 | NP_080877.3 | 6 | 770 | ||
| GenBank mRNA | AK004328.1 | - | 5 | 358 | ||
| AK020099.1 | - | 6 | 644 | |||
| BC024619.1 | - | 6 | 758 | 4923664 | ||
| BC100401.1 | - | 6 | 734 | |||
| BX525112.1 | - | 4 | 211 | |||
| CB952524.1 | - | 4 | 275 | |||

Hyi
Genomic Map