siRNA Details
| Gene Symbol: Hyi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026601.3 | NP_080877.3 | 4, 5 | 577 | ||
| GenBank mRNA | AK004328.1 | - | 3, 4 | 165 | ||
| AK020099.1 | - | 4, 5 | 451 | |||
| BC024619.1 | - | 4, 5 | 565 | 4923664 | ||
| BC100401.1 | - | 4, 5 | 541 | |||
| CB952524.1 | - | 2, 3 | 82 | |||
| CK620463.1 | - | 4, 5 | 465 | |||

Hyi
Genomic Map