siRNA Details
| Gene Symbol: AI846148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033139.3 | NP_001028311.2 | 3 | 755 | ||
| NM_001167831.1 | NP_001161303.1 | 3 | 755 | |||
| GenBank mRNA | AK090385.1 | - | 3 | 724 | ||
| AK142757.1 | - | 2 | 150 | |||
| AK163590.1 | - | 3 | 556 | |||
| AK166227.1 | - | 3 | 725 | |||
| AK168010.1 | - | 3 | 651 | |||
| BC125249.1 | - | 3 | 583 | 40059530 | ||

AI846148
Genomic Map