siRNA Details
| Gene Symbol: AI846148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033139.3 | NP_001028311.2 | 6 | 1315 | ||
| GenBank mRNA | AK090385.1 | - | 6 | 1284 | ||
| AK142757.1 | - | 5 | 711 | |||
| AK163590.1 | - | 6 | 1116 | |||
| AK166227.1 | - | 6 | 1285 | |||
| BC125249.1 | - | 6 | 1143 | 40059530 | ||

AI846148
Genomic Map