siRNA Details
| Gene Symbol: Ggct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026637.3 | NP_080913.1 | 3 | 391 | ||
| GenBank mRNA | AK008719.1 | - | 3 | 396 | ||
| AK028830.1 | - | 393 | ||||
| AK050049.1 | - | 3 | 476 | |||
| AK079465.1 | - | 3 | 393 | |||
| AK141474.1 | - | 441 | ||||
| BC080818.1 | - | 3 | 371 | 30286931 | ||

Ggct
Genomic Map