siRNA Details
| Gene Symbol: Sirt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178848.3 | NP_849179.1 | 3, 4 | 520 | ||
| GenBank mRNA | AK002609.1 | - | 4, 5 | 550 | ||
| BC031770.1 | - | 3, 4 | 520 | 4014119 | ||
| BC087898.1 | - | 4, 5 | 542 | 30254686 | ||

Sirt5
Genomic Map