siRNA Details
| Gene Symbol: Sirt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178848.3 | NP_849179.1 | 8 | 958 | ||
| GenBank mRNA | AK002609.1 | - | 9 | 987 | ||
| BC031770.1 | - | 8 | 958 | 4014119 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sirt5| Gene Symbol: Sirt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178848.3 | NP_849179.1 | 8 | 958 | ||
| GenBank mRNA | AK002609.1 | - | 9 | 987 | ||
| BC031770.1 | - | 8 | 958 | 4014119 | ||