siRNA Details
| Gene Symbol: Mob1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026735.2 | NP_081011.1 | 6 | 851 | ||
| GenBank mRNA | AK077320.1 | - | 6 | 827 | ||
| AK150016.1 | - | 6 | 844 | |||
| AK162120.1 | - | 6 | 851 | |||
| AK172291.1 | - | 6 | 819 | |||
| BC104402.1 | - | 7 | 682 | 40050502 | ||
| BC104403.1 | - | 6 | 746 | 40050503 | ||

Mob1b
Genomic Map