siRNA Details
| Gene Symbol: Mob1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026735.2 | NP_081011.1 | 5 | 763 | ||
| GenBank mRNA | AK077320.1 | - | 5 | 739 | ||
| AK150016.1 | - | 5 | 756 | |||
| AK162120.1 | - | 5 | 763 | |||
| AK172291.1 | - | 5 | 731 | |||
| BC104402.1 | - | 6 | 594 | 40050502 | ||
| BC104403.1 | - | 5 | 658 | 40050503 | ||

Mob1b
Genomic Map