siRNA Details
Gene Symbol: Arel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178065.4 | NP_835166.3 | 18 | 2809 | ||
GenBank mRNA | AB093230.1 | - | 17 | 2428 | ||
AK077015.1 | - | 17 | 2341 | |||
AK133332.1 | - | 17 | 2345 | |||
AK147491.1 | - | 18 | 5024 | |||
AK147878.1 | - | 18 | 2773 | |||
AK154529.1 | - | 18 | 2790 | |||
BC006074.1 | - | 9 | 964 | |||
BC049900.1 | - | 12 | 1499 | |||
BC057351.1 | - | 17 | 2637 | |||
BC060658.1 | - | 17 | 2378 | 30355406 | ||
BC079590.1 | - | 18 | 2763 |
