siRNA Details
Gene Symbol: Pmvk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026784.3 | NP_081060.2 | 3 | 473 | ||
NM_027348.1 | NP_081624.1 | 3 | 442 | |||
GenBank mRNA | AK003607.1 | - | 3 | 276 | ||
AK013477.1 | - | 3 | 442 | |||
BC028659.1 | - | 2 | 261 | 1247653 |
