siRNA Details
| Gene Symbol: Mybphl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026831.1 | NP_081107.1 | 7, 8 | 1140 | ||
| GenBank mRNA | AK004148.1 | - | 7, 8 | 1141 | ||
| BC089625.1 | - | 7, 8 | 1128 | 6756382 | ||
| BC132636.1 | - | 7, 8 | 1129 | 40130913 | ||
| BC132638.1 | - | 7, 8 | 1129 | 40130915 | ||

Mybphl
Genomic Map