siRNA Details
| Gene Symbol: Cgrrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026832.3 | NP_081108.2 | 1, 2 | 146 | ||
| GenBank mRNA | AK004156.1 | - | 1, 2 | 172 | ||
| AK007402.1 | - | 1, 2 | 162 | |||
| AK030403.1 | - | 1, 2 | 147 | |||
| AK030755.1 | - | 1, 2 | 148 | |||
| AK154797.1 | - | 1, 2 | 222 | |||
| BC037677.1 | - | 1, 2 | 165 | 3491666 | ||

Cgrrf1
Genomic Map